siRNA Details
| Gene Symbol: Bpifb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077680.1 | NP_001071148.2 | 13, 14 | 1295 | ||
| XM_006235355.2 | XP_006235417.1 | 13, 14 | 1374 | |||
| GenBank mRNA | BC127515.1 | - | 13, 14 | 1295 | 7100079 | |

Bpifb1
Genomic Map