siRNA Details
| Gene Symbol: Galntl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025148.1 | NP_001020319.1 | 8 | 1205 | ||
| XM_006235958.2 | XP_006236020.1 | 7 | 1168 | |||
| GenBank mRNA | BC078995.1 | - | 8 | 1205 | 7112721 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galntl5| Gene Symbol: Galntl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025148.1 | NP_001020319.1 | 8 | 1205 | ||
| XM_006235958.2 | XP_006236020.1 | 7 | 1168 | |||
| GenBank mRNA | BC078995.1 | - | 8 | 1205 | 7112721 | |