siRNA Details
| Gene Symbol: LOC499971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024317.2 | NP_001019488.2 | 17 | 2038 | ||
| XM_008762655.1 | XP_008760877.1 | 16 | 1691 | |||
| XM_008762656.1 | XP_008760878.1 | 15 | 1582 | |||
| GenBank mRNA | BC079392.1 | - | 10 | 974 | 7111961 | |

LOC499971
Genomic Map