siRNA Details
| Gene Symbol: Atg9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191968.1 | NP_001178897.1 | 1 | 260 | ||
| XM_006235960.2 | XP_006236022.1 | 1 | 260 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atg9b| Gene Symbol: Atg9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191968.1 | NP_001178897.1 | 1 | 260 | ||
| XM_006235960.2 | XP_006236022.1 | 1 | 260 | |||