siRNA Details
| Gene Symbol: Nupl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109217.1 | NP_001102687.1 | 7 | 1016 | ||
| GenBank mRNA | FQ227080.1 | - | 7 | 983 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nupl2| Gene Symbol: Nupl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109217.1 | NP_001102687.1 | 7 | 1016 | ||
| GenBank mRNA | FQ227080.1 | - | 7 | 983 | ||