siRNA Details
| Gene Symbol: Fam126a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191969.1 | NP_001178898.1 | 9, 10 | 1068 | ||
| XM_006235961.2 | XP_006236023.1 | 9, 10 | 1370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam126a| Gene Symbol: Fam126a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191969.1 | NP_001178898.1 | 9, 10 | 1068 | ||
| XM_006235961.2 | XP_006236023.1 | 9, 10 | 1370 | |||