siRNA Details
| Gene Symbol: RGD1564345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077681.1 | NP_001071149.1 | 2 | 170 | ||
| GenBank mRNA | BC127528.1 | - | 2 | 170 | 7114751 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564345| Gene Symbol: RGD1564345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077681.1 | NP_001071149.1 | 2 | 170 | ||
| GenBank mRNA | BC127528.1 | - | 2 | 170 | 7114751 | |