siRNA Details
| Gene Symbol: Cldn12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100813.1 | NP_001094283.1 | 2 | 301 | ||
| XM_006236052.2 | XP_006236114.1 | 4 | 526 | |||
| XM_006236053.2 | XP_006236115.1 | 4 | 508 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn12| Gene Symbol: Cldn12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100813.1 | NP_001094283.1 | 2 | 301 | ||
| XM_006236052.2 | XP_006236114.1 | 4 | 526 | |||
| XM_006236053.2 | XP_006236115.1 | 4 | 508 | |||