siRNA Details
| Gene Symbol: RGD1563091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236079.2 | XP_006236141.1 | 2 | 2651 | ||
| XM_575365.5 | XP_575365.2 | 3 | 2916 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1563091| Gene Symbol: RGD1563091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236079.2 | XP_006236141.1 | 2 | 2651 | ||
| XM_575365.5 | XP_575365.2 | 3 | 2916 | |||