siRNA Details
| Gene Symbol: Asb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024318.1 | NP_001019489.1 | 1 | 120 | ||
| XM_008762714.1 | XP_008760936.1 | 1 | 412 | |||
| GenBank mRNA | AY577765.1 | - | 1 | 120 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb4| Gene Symbol: Asb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024318.1 | NP_001019489.1 | 1 | 120 | ||
| XM_008762714.1 | XP_008760936.1 | 1 | 412 | |||
| GenBank mRNA | AY577765.1 | - | 1 | 120 | ||