siRNA Details
| Gene Symbol: Dlx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762741.1 | XP_008760963.1 | 2 | 870 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlx6| Gene Symbol: Dlx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762741.1 | XP_008760963.1 | 2 | 870 | ||