siRNA Details
| Gene Symbol: LOC500028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047954.1 | NP_001041419.1 | 1 | 622 | ||
| GenBank mRNA | AY383683.1 | - | 1 | 622 | ||
| Gene Symbol: Yae1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107356.2 | NP_001100826.1 | 3 | 623 | ||
| GenBank mRNA | FQ211942.1 | - | 3 | 629 | ||
| FQ216975.1 | - | 3 | 590 | |||

LOC500028
Genomic Map