siRNA Details
| Gene Symbol: Phf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110492.1 | NP_001103962.1 | 8 | 1878 | ||
| XM_006236104.2 | XP_006236166.1 | 8 | 1914 | |||
| XM_006236105.2 | XP_006236167.1 | 8 | 1653 | |||
| XM_006236107.2 | XP_006236169.1 | 8 | 1914 | |||
| XM_006236108.2 | XP_006236170.1 | 8 | 1914 | |||
| XM_006236109.2 | XP_006236171.1 | 8 | 1914 | |||
| XM_006236111.2 | XP_006236173.1 | 8 | 1913 | |||
| XM_008762716.1 | XP_008760938.1 | 8 | 1914 | |||
| GenBank mRNA | BC166456.1 | - | 8 | 1609 | 9025134 | |
| FQ221829.1 | - | 8 | 1843 | |||

Phf14
Genomic Map