siRNA Details
| Gene Symbol: Wnt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109223.1 | NP_001102693.1 | 3 | 849 | ||
| XM_006236143.2 | XP_006236205.1 | 3 | 898 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt16| Gene Symbol: Wnt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109223.1 | NP_001102693.1 | 3 | 849 | ||
| XM_006236143.2 | XP_006236205.1 | 3 | 898 | |||