siRNA Details
| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109224.1 | NP_001102694.1 | 4 | 1517 | ||
| XM_008762805.1 | XP_008761027.1 | 5 | 1079 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fezf1| Gene Symbol: Fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109224.1 | NP_001102694.1 | 4 | 1517 | ||
| XM_008762805.1 | XP_008761027.1 | 5 | 1079 | |||