siRNA Details
| Gene Symbol: Prrt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109226.1 | NP_001102696.1 | 2 | 355 | ||
| XM_006236220.2 | XP_006236282.1 | 2 | 562 | |||
| XM_008762817.1 | XP_008761039.1 | 2 | 246 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prrt4| Gene Symbol: Prrt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109226.1 | NP_001102696.1 | 2 | 355 | ||
| XM_006236220.2 | XP_006236282.1 | 2 | 562 | |||
| XM_008762817.1 | XP_008761039.1 | 2 | 246 | |||