siRNA Details
| Gene Symbol: Fam71f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024324.1 | NP_001019495.1 | 3 | 555 | ||
| XM_006236223.2 | XP_006236285.1 | 5 | 917 | |||
| XM_006236224.2 | XP_006236286.1 | 5 | 917 | |||
| XM_006236225.2 | XP_006236287.1 | 5 | 917 | |||
| XM_008762818.1 | XP_008761040.1 | 5 | 917 | |||
| GenBank mRNA | BC079297.1 | - | 3 | 555 | 7134497 | |

Fam71f1
Genomic Map