siRNA Details
| Gene Symbol: Cep41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025770.2 | NP_001020941.1 | 2 | 149 | ||
| XM_006236229.2 | XP_006236291.1 | 4 | 324 | |||
| GenBank mRNA | BC098832.1 | - | 2 | 151 | 7378587 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep41| Gene Symbol: Cep41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025770.2 | NP_001020941.1 | 2 | 149 | ||
| XM_006236229.2 | XP_006236291.1 | 4 | 324 | |||
| GenBank mRNA | BC098832.1 | - | 2 | 151 | 7378587 | |