siRNA Details
| Gene Symbol: UBE4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004788.3 | NP_004779.2 | 6 | 864 | ||
| GenBank mRNA | D50916.1 | - | 6 | 801 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBE4A| Gene Symbol: UBE4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004788.3 | NP_004779.2 | 6 | 864 | ||
| GenBank mRNA | D50916.1 | - | 6 | 801 | ||