siRNA Details
| Gene Symbol: Stra8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236282.2 | XP_006236344.1 | 8 | 3391 | ||
| XM_575429.4 | XP_575429.2 | 7 | 1134 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stra8| Gene Symbol: Stra8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236282.2 | XP_006236344.1 | 8 | 3391 | ||
| XM_575429.4 | XP_575429.2 | 7 | 1134 | |||