siRNA Details
Gene Symbol: Trim24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044266.1 | NP_001037731.1 | 12, 13 | 1693 | ||
GenBank mRNA | AY325210.1 | - | 12, 13 | 1693 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044266.1 | NP_001037731.1 | 12, 13 | 1693 | ||
GenBank mRNA | AY325210.1 | - | 12, 13 | 1693 |