siRNA Details
| Gene Symbol: Ttc26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025045.1 | NP_001020216.1 | 3, 4 | 339 | ||
| XM_006236330.2 | XP_006236392.1 | 3, 4 | 370 | |||
| GenBank mRNA | BC085935.1 | - | 3, 4 | 339 | 7115235 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc26| Gene Symbol: Ttc26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025045.1 | NP_001020216.1 | 3, 4 | 339 | ||
| XM_006236330.2 | XP_006236392.1 | 3, 4 | 370 | |||
| GenBank mRNA | BC085935.1 | - | 3, 4 | 339 | 7115235 | |