siRNA Details
| Gene Symbol: Fmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037795.1 | NP_001032884.1 | 2 | 278 | ||
| GenBank mRNA | BC098843.1 | - | 2 | 278 | 7379283 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fmc1| Gene Symbol: Fmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037795.1 | NP_001032884.1 | 2 | 278 | ||
| GenBank mRNA | BC098843.1 | - | 2 | 278 | 7379283 | |