siRNA Details
| Gene Symbol: Tas2r137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025149.1 | NP_001020320.1 | 1 | 244 | ||
| XM_006236362.2 | XP_006236424.1 | 2 | 6234 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r137| Gene Symbol: Tas2r137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025149.1 | NP_001020320.1 | 1 | 244 | ||
| XM_006236362.2 | XP_006236424.1 | 2 | 6234 | |||