siRNA Details
Gene Symbol: Tas2r138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024685.1 | NP_001019856.1 | 1 | 477 | ||
GenBank mRNA | AY916512.1 | - | 1 | 477 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tas2r138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024685.1 | NP_001019856.1 | 1 | 477 | ||
GenBank mRNA | AY916512.1 | - | 1 | 477 |