siRNA Details
| Gene Symbol: Lrrc61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109231.1 | NP_001102701.1 | 3 | 534 | ||
| XM_006236435.2 | XP_006236497.1 | 2 | 1551 | |||
| XM_006236436.2 | XP_006236498.1 | 2 | 402 | |||
| XM_006236437.2 | XP_006236499.1 | 2 | 569 | |||
| XM_006236438.2 | XP_006236500.1 | 2 | 254 | |||

Lrrc61
Genomic Map