siRNA Details
| Gene Symbol: RGD1563352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_575471.4 | XP_575471.1 | 1 | 108 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1563352| Gene Symbol: RGD1563352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_575471.4 | XP_575471.1 | 1 | 108 | ||