siRNA Details
| Gene Symbol: LOC100910298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749763.2 | XP_003749811.1 | 2 | 653 | ||
| Gene Symbol: Hoxa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271355.1 | NP_001258284.1 | 2 | 1042 | ||
| Gene Symbol: LOC103692142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762959.1 | XP_008761181.1 | 3 | 437 | ||

LOC100910298
Genomic Map