siRNA Details
| Gene Symbol: Evx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191972.1 | NP_001178901.1 | 1 | 69 | ||
| Gene Symbol: NEWGENE_1565788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762952.1 | XP_008761174.1 | 1 | 80 | ||

Evx1
Genomic Map