siRNA Details
| Gene Symbol: Nod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109236.1 | NP_001102706.1 | 4 | 2420 | ||
| XM_008762910.1 | XP_008761132.1 | 4 | 2694 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nod1| Gene Symbol: Nod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109236.1 | NP_001102706.1 | 4 | 2420 | ||
| XM_008762910.1 | XP_008761132.1 | 4 | 2694 | |||