siRNA Details
| Gene Symbol: Neurod6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109237.1 | NP_001102707.1 | 2 | 866 | ||
| GenBank mRNA | BC168855.1 | - | 1 | 892 | 8365463 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Neurod6| Gene Symbol: Neurod6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109237.1 | NP_001102707.1 | 2 | 866 | ||
| GenBank mRNA | BC168855.1 | - | 1 | 892 | 8365463 | |