siRNA Details
| Gene Symbol: Ccdc129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191973.1 | NP_001178902.1 | 10 | 1346 | ||
| XM_008762911.1 | XP_008761133.1 | 10 | 1363 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc129| Gene Symbol: Ccdc129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191973.1 | NP_001178902.1 | 10 | 1346 | ||
| XM_008762911.1 | XP_008761133.1 | 10 | 1363 | |||