siRNA Details
Gene Symbol: Mmrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001071128.5 | XP_001071128.2 | 8 | 3522 | ||
XM_006236596.2 | XP_006236658.1 | 8 | 3519 | |||
XM_006236597.2 | XP_006236659.1 | 9 | 3520 |
