siRNA Details
| Gene Symbol: RGD1560028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109239.1 | NP_001102709.1 | 5, 6 | 1050 | ||
More Information
Other Species Targeted
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560028| Gene Symbol: RGD1560028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109239.1 | NP_001102709.1 | 5, 6 | 1050 | ||
Set Membership: |
|