siRNA Details
| Gene Symbol: Ctnna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106598.1 | NP_001100068.1 | 19 | 2904 | ||
| XM_008763020.1 | XP_008761242.1 | 10 | 1941 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctnna2| Gene Symbol: Ctnna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106598.1 | NP_001100068.1 | 19 | 2904 | ||
| XM_008763020.1 | XP_008761242.1 | 10 | 1941 | |||