siRNA Details
| Gene Symbol: Lrrtm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134746.1 | NP_001128218.1 | 1 | 1498 | ||
| XM_008763024.1 | XP_008761246.1 | 2 | 1817 | |||
| XM_008763025.1 | XP_008761247.1 | 3 | 1970 | |||
| XM_008763026.1 | XP_008761248.1 | 3 | 1881 | |||
| GenBank mRNA | BC168219.1 | - | 1 | 1498 | 7319359 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_178731, NM_001282102, NM_001134743, AK032470, AK038656, and others.

Lrrtm4
Genomic Map