siRNA Details
| Gene Symbol: Emx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_575584.5 | XP_575584.5 | 3 | 1101 | ||
| GenBank mRNA | FQ212435.1 | - | 4 | 2308 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emx1| Gene Symbol: Emx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_575584.5 | XP_575584.5 | 3 | 1101 | ||
| GenBank mRNA | FQ212435.1 | - | 4 | 2308 | ||