siRNA Details
| Gene Symbol: Cml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173449.1 | NP_001166920.1 | 1 | 637 | ||
| XM_006236785.2 | XP_006236847.1 | 3 | 998 | |||
| XM_006236786.2 | XP_006236848.1 | 3 | 1024 | |||
| XM_006236787.2 | XP_006236849.1 | 3 | 1008 | |||
| Gene Symbol: LOC100910829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236799.2 | XP_006236861.1 | 3 | 909 | ||
| XM_006236800.2 | XP_006236862.1 | 908 | ||||
| XM_006236801.2 | XP_006236863.1 | 894 | ||||

Cml2
Genomic Map