siRNA Details
| Gene Symbol: CPNE6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001280558.1 | NP_001267487.1 | 13, 14 | 1425 | ||
| NM_006032.3 | NP_006023.1 | 12, 13 | 1413 | |||
| XM_005268216.1 | XP_005268273.1 | 13, 14 | 1369 | |||
| XM_005268217.1 | XP_005268274.1 | 11, 12 | 1063 | |||
| GenBank mRNA | AB009288.1 | - | 12, 13 | 1213 | ||
| AK125195.1 | - | 11, 12 | 1975 | |||
| AK222675.1 | - | 12, 13 | 1173 | |||
| AK293655.1 | - | 13, 14 | 1425 | |||
| AK314182.1 | - | 12, 13 | 1226 | |||
| AL137688.1 | - | 5, 6 | 3420 | |||
| BC018046.1 | - | 12, 13 | 1219 | 4799650 | ||

CPNE6
Genomic Map