siRNA Details
| Gene Symbol: Bmp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031824.1 | NP_001026994.1 | 2 | 907 | ||
| GenBank mRNA | AB158417.1 | - | 2 | 907 | ||
| AB158418.1 | - | 2 | 907 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmp10| Gene Symbol: Bmp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031824.1 | NP_001026994.1 | 2 | 907 | ||
| GenBank mRNA | AB158417.1 | - | 2 | 907 | ||
| AB158418.1 | - | 2 | 907 | |||