siRNA Details
| Gene Symbol: Hmces | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025047.1 | NP_001020218.1 | 2 | 324 | ||
| XM_006236851.2 | XP_006236913.1 | 3 | 708 | |||
| GenBank mRNA | BC083690.1 | - | 2 | 324 | 7189409 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmces| Gene Symbol: Hmces | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025047.1 | NP_001020218.1 | 2 | 324 | ||
| XM_006236851.2 | XP_006236913.1 | 3 | 708 | |||
| GenBank mRNA | BC083690.1 | - | 2 | 324 | 7189409 | |