siRNA Details
| Gene Symbol: Dnajb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109248.1 | NP_001102718.1 | 1 | 815 | ||
| XM_008763077.1 | XP_008761299.1 | 1 | 850 | |||
| XM_008763078.1 | XP_008761300.1 | 1 | 850 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajb8| Gene Symbol: Dnajb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109248.1 | NP_001102718.1 | 1 | 815 | ||
| XM_008763077.1 | XP_008761299.1 | 1 | 850 | |||
| XM_008763078.1 | XP_008761300.1 | 1 | 850 | |||