siRNA Details
| Gene Symbol: RAB33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004794.2 | NP_004785.1 | 1 | 371 | ||
| GenBank mRNA | AF498959.1 | - | 1 | 107 | ||
| BC001157.1 | - | 1 | 371 | 3510745 | ||
| BC009996.2 | - | 1 | 371 | 4129324 | ||
| BT007038.1 | - | 1 | 107 | |||
| D14889.1 | - | 1 | 544 | |||

RAB33A
Genomic Map