siRNA Details
Gene Symbol: Lrrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037363.1 | NP_001032440.1 | 1 | 642 | ||
XM_006236984.2 | XP_006237046.1 | 2 | 1353 | |||
XM_006236985.2 | XP_006237047.1 | 2 | 1920 | |||
GenBank mRNA | BC107902.1 | - | 1 | 642 | 7109029 |
