siRNA Details
| Gene Symbol: Setmar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025048.1 | NP_001020219.1 | 1, 2 | 185 | ||
| XM_008763195.1 | XP_008761417.1 | 1, 2 | 219 | |||
| GenBank mRNA | BC088181.1 | - | 1, 2 | 185 | 7364937 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Setmar| Gene Symbol: Setmar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025048.1 | NP_001020219.1 | 1, 2 | 185 | ||
| XM_008763195.1 | XP_008761417.1 | 1, 2 | 219 | |||
| GenBank mRNA | BC088181.1 | - | 1, 2 | 185 | 7364937 | |