siRNA Details
| Gene Symbol: Setmar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025048.1 | NP_001020219.1 | 2 | 248 | ||
| XM_006236996.2 | XP_006237058.1 | 2 | 119 | |||
| XM_006236997.2 | XP_006237059.1 | 2 | 343 | |||
| XM_008763195.1 | XP_008761417.1 | 2 | 282 | |||
| GenBank mRNA | BC088181.1 | - | 2 | 248 | 7364937 | |

Setmar
Genomic Map