siRNA Details
| Gene Symbol: Cecr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191977.1 | NP_001178906.1 | 17 | 3521 | ||
| XM_008763251.1 | XP_008761473.1 | 17 | 3881 | |||
| XM_008763252.1 | XP_008761474.1 | 17 | 3878 | |||
| XM_008763253.1 | XP_008761475.1 | 16 | 3797 | |||

Cecr2
Genomic Map