siRNA Details
| Gene Symbol: Cecr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191977.1 | NP_001178906.1 | 16 | 2245 | ||
| XM_008763251.1 | XP_008761473.1 | 16 | 2605 | |||
| XM_008763252.1 | XP_008761474.1 | 16 | 2605 | |||
| XM_008763253.1 | XP_008761475.1 | 15 | 2521 | |||

Cecr2
Genomic Map