siRNA Details
| Gene Symbol: Cd27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024335.1 | NP_001019506.1 | 2 | 274 | ||
| XM_006237380.2 | XP_006237442.1 | 2 | 1784 | |||
| XM_006237381.2 | XP_006237443.1 | 2 | 1784 | |||
| XM_006237382.2 | XP_006237444.1 | 2 | 1784 | |||
| XM_008763293.1 | XP_008761515.1 | 3 | 1579 | |||
| GenBank mRNA | BC095844.1 | - | 2 | 274 | 7389613 | |

Cd27
Genomic Map