siRNA Details
| Gene Symbol: Cracr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237505.2 | XP_006237567.1 | 12 | 3281 | ||
| XM_006237508.2 | XP_006237570.1 | 12 | 3281 | |||
| XM_008763306.1 | XP_008761528.1 | 12 | 3281 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cracr2a| Gene Symbol: Cracr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237505.2 | XP_006237567.1 | 12 | 3281 | ||
| XM_006237508.2 | XP_006237570.1 | 12 | 3281 | |||
| XM_008763306.1 | XP_008761528.1 | 12 | 3281 | |||